Gene Transcripts

Gene Symbol
Lrrtm4
Entrez Gene ID
243499
Gene Name
leucine rich repeat transmembrane neuronal 4
Gene Aliases
7530419J18Rik, A230052N11
Chromosome Location
Chr. 6: 80018859 - 80810162 on Build GRCm38
UniGene ID
Mm.483425
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001282102.1 NP_001269031.1 2 1865
NM_178731.5 NP_848846.3 2 1619
GenBank mRNA AK038656.1 - 2 1696

View Full Product Details