Gene Transcripts

Gene Symbol
Ndufb9
Entrez Gene ID
66218
Gene Name
NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 9
Gene Aliases
1190008J14Rik
Chromosome Location
Chr. 15: 58933810 - 58939489 on Build GRCm38
UniGene ID
Mm.322294
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023172.3 NP_075661.1 1, 2 137
GenBank mRNA AK002254.1 - 1, 2 138
AK003540.1 - 1, 2 138
AK004522.1 - 1, 2 138
BC031539.1 - 1, 2 119 3979845

View Full Product Details