Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_027514.2 | NP_081790.1 | 2 | 324 | |
GenBank mRNA | AB195430.1 | - | 2 | 281 | |
AJ563420.1 | - | 2 | 212 | ||
AK014446.1 | - | 2 | 295 | ||
AK038747.1 | - | 2 | 295 | ||
AK040334.1 | - | 2 | 295 | ||
AK078344.1 | - | 2 | 291 | ||
AK080753.1 | - | 2 | 295 | ||
AK165873.1 | - | 2 | 314 | ||
BC013673.1 | - | 2 | 281 | 4188634 | |
BC032283.1 | - | 2 | 284 | 5363187 | |
BC138219.1 | - | 2 | 269 | 8861238 | |
BC138220.1 | - | 2 | 269 | 8861239 | |
BY276828.1 | - | 2 | 325 |