Gene Transcripts

Gene Symbol
Dpy30
Entrez Gene ID
66310
Gene Name
dpy-30 homolog (C. elegans)
Gene Aliases
2810410M20Rik, C87842
Chromosome Location
Chr. 17: 74299474 - 74323944 on Build GRCm38
UniGene ID
Mm.28536
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001146222.1 NP_001139694.1 4, 5 442
NM_001146223.1 NP_001139695.1 4, 5 356
NM_001146224.1 NP_001139696.1 3, 4 362
NM_024428.4 NP_077746.3 4, 5 451
GenBank mRNA AK003688.1 - 3, 4 287
AK011643.1 - 3, 4 261
AK013073.1 - 4, 5 345
AK077902.1 - 4, 5 344
AK214978.1 - 3, 4 234
AV460563.1 - 4, 5 363
AY297538.1 - 4, 5 455
BC002240.1 - 4, 5 325 3491912
BU961422.1 - 4, 5 457
BX518196.1 - 4, 5 457

View Full Product Details