Gene Transcripts

Gene Symbol
Ndufb4
Entrez Gene ID
68194
Gene Name
NADH dehydrogenase (ubiquinone) 1 beta subcomplex 4
Gene Aliases
0610006N12Rik, 1300010H20Rik
Chromosome Location
Chr. 16: 37647602 - 37654368 on Build GRCm38
UniGene ID
Mm.379154
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026610.1 NP_080886.1 3 372
GenBank mRNA AK009807.1 - 3 372
AK018707.1 - 3 372
BC056229.1 - 3 373 6475281
BC096421.1 - 3 371

View Full Product Details