Gene Transcripts

Gene Symbol
Acyp2
Entrez Gene ID
75572
Gene Name
acylphosphatase 2, muscle type
Gene Aliases
2310004B09Rik
Chromosome Location
Chr. 11: 30505992 - 30649396 on Build GRCm38
UniGene ID
Mm.390319
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029344.3 NP_083620.1 3, 4 220
GenBank mRNA AK009134.1 - 3, 4 220
AK046238.1 - 3, 4 229
BC027642.1 - 3, 4 208 1245384

View Full Product Details