Gene Transcripts

Gene Symbol
G0s2
Entrez Gene ID
14373
Gene Name
G0/G1 switch gene 2
Gene Aliases
AI255151, AV006465
Chromosome Location
Chr. 1: 193272160 - 193273188 on Build GRCm38
UniGene ID
Mm.439721
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008059.3 NP_032085.1 2 566
GenBank mRNA AK003165.1 - 2 488
BC117038.1 - 2 394 40126357
BC117040.1 - 2 394 40126359
X95280.1 - 2 470

View Full Product Details