Gene Transcripts

Gene Symbol
Krtap21-1
Entrez Gene ID
170656
Gene Name
keratin associated protein 21-1
Gene Aliases
1110017B05Rik, AI506979, KAP21.1, Krtap16-7
Chromosome Location
Chr. 16: 89403027 - 89403774 on Build GRCm38
UniGene ID
Mm.46109
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028621.3 NP_082897.2 1 531
GenBank mRNA AK003736.2 - 1 562
BC145974.1 - 1 531
BC145976.1 - 1 531

View Full Product Details