Gene Transcripts
- Gene Symbol
- Cd99
- Entrez Gene ID
- 673094
- Gene Name
- CD99 antigen
- Gene Aliases
- 1110061M03Rik, 2410026K10Rik, D4, Pilr-l, pilr-1
- Chromosome Location
- Chr. 4|NT_187055: 48 - 1475 on Build GRCm38
- UniGene ID
- Mm.423040
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK002762.1 |
- |
|
680 |
|
View Full Product Details