Gene Transcripts

Gene Symbol
Vmn1r33
Entrez Gene ID
171187
Gene Name
vomeronasal 1 receptor 33
Gene Aliases
V1rc14
Chromosome Location
Chr. 6: 66611651 - 66612568 on Build GRCm38
UniGene ID
Mm.451648
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_134169.1 NP_598930.1 1 549
GenBank mRNA BC138428.1 - 1 598
BC138429.1 - 1 598 8861449

View Full Product Details