Gene Transcripts

Gene Symbol
Amigo1
Entrez Gene ID
229715
Gene Name
adhesion molecule with Ig like domain 1
Gene Aliases
Amigo, ali2
Chromosome Location
Chr. 3: 108186289 - 108192286 on Build GRCm38
UniGene ID
Mm.275752
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004293.2 NP_001004293.1 2 740
NM_001287093.1 NP_001274022.1 2 740
NM_146137.3 NP_666249.2 2 775
GenBank mRNA AB167509.1 - 1 396
AK035960.1 - 2 671
AK173118.1 - 2 729
AY237008.1 - 1 289
BC010598.1 - 2 691 3498778
BC022907.1 - 2 679

View Full Product Details