Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_020264.4 | NP_064660.2 | 2 | 140 | |
GenBank mRNA | AF134204.1 | - | 2 | 101 | |
AK020540.1 | - | 2 | 124 | ||
BC120764.1 | - | 2 | 140 | 40129687 | |
BC120766.1 | - | 2 | 140 | 40129689 |