Gene Transcripts

Gene Symbol
Krtap4-2
Entrez Gene ID
68673
Gene Name
keratin associated protein 4-2
Gene Aliases
1110033F04Rik, AI604442
Chromosome Location
Chr. 11: 99634114 - 99635084 on Build GRCm38
UniGene ID
Mm.351893
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026807.2 NP_081083.2 1 343
GenBank mRNA AK004055.1 - 1 344

View Full Product Details