Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001290624.1 | NP_001277553.1 | 3, 4 | 397 | |
GenBank mRNA | AK037217.1 | - | 3, 4 | 360 | |
AK083704.1 | - | 2, 3 | 364 | ||
BC023671.1 | - | 3, 4 | 393 | ||
BC049258.1 | - | 3, 4 | 339 | 5250488 |