Gene Transcripts

Gene Symbol
Aqp11
Entrez Gene ID
66333
Gene Name
aquaporin 11
Gene Aliases
1700015P13Rik, AI648923, sjds
Chromosome Location
Chr. 7: 97725490 - 97738514 on Build GRCm38
UniGene ID
Mm.29756
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175105.3 NP_780314.1 1 418
GenBank mRNA AB028148.1 - 1 172
AK006005.1 - 1 444
AK040045.1 - 1 187
AK075757.1 - 1 419
AK137326.1 - 1 461
BC037088.1 - 1 197
BC068103.1 - 2 67

View Full Product Details