Gene Transcripts
- Gene Symbol
- MAPK11
- Entrez Gene ID
- 5600
- Gene Name
- mitogen-activated protein kinase 11
- Gene Aliases
- P38B, P38BETA2, PRKM11, SAPK2, SAPK2B, p38-2, p38Beta
- Chromosome Location
- Chr. 22: 50263713 - 50270393 on Build GRCh38
- UniGene ID
- Hs.57732
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_002751.6 |
NP_002742.3
|
3, 4 |
408 |
|
NR_110887.1 |
|
3, 4 |
408 |
|
GenBank mRNA |
AF001008.1 |
- |
3, 4 |
307 |
|
AF001174.1 |
- |
3, 4 |
313 |
|
AF031135.1 |
- |
3, 4 |
322 |
|
AK291845.1 |
- |
3, 4 |
359 |
|
AK308969.1 |
- |
3, 4 |
324 |
|
AK309046.1 |
- |
3, 4 |
510 |
|
BC027933.1 |
- |
3, 4 |
365 |
5199628 |
BM857051.1 |
- |
3, 4 |
392 |
|
BX361670.2 |
- |
3, 4 |
392 |
|
BX444192.2 |
- |
3, 4 |
407 |
|
CR456514.1 |
- |
3, 4 |
309 |
|
U53442.1 |
- |
3, 4 |
326 |
|
U92268.1 |
- |
3, 4 |
311 |
|
Y14440.1 |
- |
3, 4 |
313 |
|
View Full Product Details