Gene Transcripts

Gene Symbol
PPM1D
Entrez Gene ID
8493
Gene Name
protein phosphatase, Mg2+/Mn2+ dependent 1D
Gene Aliases
PP2C-DELTA, WIP1
Chromosome Location
Chr. 17: 60600183 - 60666280 on Build GRCh38
UniGene ID
Hs.286073
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003620.3 NP_003611.1 4 1239
GenBank mRNA AK299773.1 - 5 948
AK312902.1 - 4 1229
BC016480.1 - 4 1155 3923593
BC033893.2 - 4 1113 5265521
BC042418.1 - 4 1153 5167004
BC060877.1 - 4 1239 5260081
BT009780.1 - 4 1007
KU178420.1 - 4 1007
KU178421.1 - 3 778
U78305.1 - 4 1210

View Full Product Details