Gene Transcripts

Gene Symbol
MTMR1
Entrez Gene ID
8776
Gene Name
myotubularin related protein 1
Gene Aliases
Chromosome Location
Chr. X: 150692997 - 150765103 on Build GRCh38
UniGene ID
Hs.347187
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001306145.1 NP_001293074.1 2, 3 427
NM_003828.3 NP_003819.1 2, 3 427
XM_006724855.3 XP_006724918.1 2, 3 427
XM_011531209.2 XP_011529511.1 2, 3 187
GenBank mRNA AF057354.1 - 2, 3 251
AJ224979.1 - 2, 3 253
BC032947.1 - 2, 3 424 4837744
DR000536.1 - 2, 3 429

View Full Product Details