Gene Transcripts

Gene Symbol
CTSZ
Entrez Gene ID
1522
Gene Name
cathepsin Z
Gene Aliases
CTSX
Chromosome Location
Chr. 20: 58995187 - 59007254 on Build GRCh38
UniGene ID
Hs.252549
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001336.3 NP_001327.2 6 1452
GenBank mRNA AF009923.1 - 6 1295
AF136273.1 - 6 1451
BC042168.1 - 6 1435 5018854

View Full Product Details