Gene Transcripts

Gene Symbol
HMCN1
Entrez Gene ID
83872
Gene Name
hemicentin 1
Gene Aliases
ARMD1, FBLN6, FIBL-6, FIBL6
Chromosome Location
Chr. 1: 185734526 - 186190953 on Build GRCh38
UniGene ID
Hs.58877
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031935.2 NP_114141.2 48 7707
XM_011510038.2 XP_011508340.1 48 7724
XM_011510041.2 XP_011508343.1 48 7732
XM_017002437.1 XP_016857926.1 37 5657
GenBank mRNA AF156100.1 - 48 7710

View Full Product Details