Gene Transcripts

Gene Symbol
PSMB4
Entrez Gene ID
5692
Gene Name
proteasome subunit beta 4
Gene Aliases
HN3, HsN3, PROS-26, PROS26
Chromosome Location
Chr. 1: 151399565 - 151401936 on Build GRCh38
UniGene ID
Hs.89545
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002796.2 NP_002787.2 3 425
GenBank mRNA AK294149.1 - 3 427
AK313825.1 - 3 427
BC000331.1 - 3 416
BC008314.2 - 3 424
BC010088.2 - 3 424
BC010098.2 - 3 424
BC011768.2 - 3 424
BC012168.2 - 3 424
BC017307.1 - 3 474
BC017451.2 - 3 424
BC017486.2 - 3 424
BT006917.1 - 3 402
CR456820.1 - 3 402
D26600.1 - 3 425
S71381.1 - 3 408

View Full Product Details