Gene Transcripts

Gene Symbol
CDC5L
Entrez Gene ID
988
Gene Name
cell division cycle 5 like
Gene Aliases
CDC5, CDC5-LIKE, CEF1, PCDC5RP, dJ319D22.1
Chromosome Location
Chr. 6: 44387514 - 44450424 on Build GRCh38
UniGene ID
Hs.485471
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001253.3 NP_001244.1 16 2751
GenBank mRNA AB007892.2 - 16 2523
AK128737.1 - 16 2559
AK299738.1 - 16 2478
BC001568.1 - 16 2540 3452352
BG187521.1 - 2 361

View Full Product Details