Gene Transcripts

Gene Symbol
CAMKV
Entrez Gene ID
79012
Gene Name
CaM kinase like vesicle associated
Gene Aliases
1G5, VACAMKL
Chromosome Location
Chr. 3: 49857981 - 49870222 on Build GRCh38
UniGene ID
Hs.145156
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001320147.1 NP_001307076.1 8 1156
NM_024046.4 NP_076951.2 8 1156
GenBank mRNA AK074856.1 - 8 870
AK074899.1 - 8 840
AK075294.1 - 8 851
AK295588.1 - 8 714
AK297258.1 - 8 802
AK297437.1 - 8 796
AK299953.1 - 8 799
AK315525.1 - 8 889
AL136697.1 - 7 755
AL833943.1 - 6 1214
BC000497.2 - 8 825 2820734
BC001921.1 - 8 825 2820734
BC005828.2 - 8 825 2820734
BC017363.2 - 8 827 3940018
BC019256.1 - 8 820 3502894
BX647074.1 - 7 998
CR533508.1 - 6 593

View Full Product Details