Gene Transcripts

Gene Symbol
HOXB2
Entrez Gene ID
3212
Gene Name
homeobox B2
Gene Aliases
HOX2, HOX2H, Hox-2.8, K8
Chromosome Location
Chr. 17: 48542655 - 48545031 on Build GRCh38
UniGene ID
Hs.514289
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002145.3 NP_002136.1 2 1312
XM_005257275.4 XP_005257332.1 2 881
GenBank mRNA X16665.1 - 2 1270

View Full Product Details