Gene Transcripts

Gene Symbol
NDUFA6
Entrez Gene ID
4700
Gene Name
NADH:ubiquinone oxidoreductase subunit A6
Gene Aliases
B14, CI-B14, LYRM6, NADHB14
Chromosome Location
Chr. 22: 42085526 - 42090884 on Build GRCh38
UniGene ID
Hs.274416
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002490.3 NP_002481.2 3 777
GenBank mRNA BC002772.1 - 3 638

View Full Product Details