Gene Transcripts

Gene Symbol
SLC16A1
Entrez Gene ID
6566
Gene Name
solute carrier family 16 member 1
Gene Aliases
HHF7, MCT, MCT1, MCT1D
Chromosome Location
Chr. 1: 112911847 - 112956353 on Build GRCh38
UniGene ID
Hs.75231
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001166496.1 NP_001159968.1 5 2359
NM_003051.3 NP_003042.3 5 1890
XM_011542026.2 XP_011540328.1 5 1623
XM_011542027.2 XP_011540329.1 5 1633
GenBank mRNA AJ438942.2 - 1 229
AK000641.1 - 2 775
AK296688.1 - 5 1673
AK303603.1 - 5 1732
AL162079.1 - 5 1720
BC026317.1 - 5 1733 4811581
L31801.1 - 4 1539

View Full Product Details