Gene Transcripts

Gene Symbol
CEBPE
Entrez Gene ID
1053
Gene Name
CCAAT/enhancer binding protein epsilon
Gene Aliases
C/EBP-epsilon, CRP1
Chromosome Location
Chr. 14: 23117306 - 23119611 on Build GRCh38
UniGene ID
Hs.558308
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001805.3 NP_001796.2 2 1506
GenBank mRNA BC035797.1 - 2 1089
U48866.1 - 2 925

View Full Product Details