Gene Transcripts

Gene Symbol
CRHBP
Entrez Gene ID
1393
Gene Name
corticotropin releasing hormone binding protein
Gene Aliases
CRF-BP, CRFBP
Chromosome Location
Chr. 5: 76952713 - 76981143 on Build GRCh38
UniGene ID
Hs.115617
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001882.3 NP_001873.2 4 787
GenBank mRNA AK223457.1 - 4 664
AK307729.1 - 4 597
AK311052.1 - 4 593
BC018038.1 - 4 597 4812893
CR541666.1 - 4 507
X58022.1 - 4 553

View Full Product Details