Gene Transcripts

Gene Symbol
CIR1
Entrez Gene ID
9541
Gene Name
corepressor interacting with RBPJ, 1
Gene Aliases
CIR
Chromosome Location
Chr. 2: 174348150 - 174395715 on Build GRCh38
UniGene ID
Hs.632531
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004882.3 NP_004873.3 10 1139
GenBank mRNA AF098297.1 - 10 1046
AK292389.1 - 10 1122
BC015040.1 - 11 1121
BC031018.1 - 10 1070
BC046098.2 - 10 1090 5495320
U03644.1 - 10 1081

View Full Product Details