Gene Transcripts

Gene Symbol
PLXNB3
Entrez Gene ID
5365
Gene Name
plexin B3
Gene Aliases
PLEXB3, PLEXR, PLXN6
Chromosome Location
Chr. X: 153764196 - 153779346 on Build GRCh38
UniGene ID
Hs.632833
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163257.1 NP_001156729.1 28 5010
NM_005393.2 NP_005384.2 27 4844
GenBank mRNA AB033032.1 - 27 4784
AF149019.1 - 27 4784
AK292535.1 - 27 4844
AK295762.1 - 27 4864
AK295894.1 - 26 3728
AK307894.1 - 28 4987

View Full Product Details