Gene Transcripts

Gene Symbol
BAZ1A
Entrez Gene ID
11177
Gene Name
bromodomain adjacent to zinc finger domain 1A
Gene Aliases
ACF1, WALp1, WCRF180, hACF1
Chromosome Location
Chr. 14: 34752731 - 34875647 on Build GRCh38
UniGene ID
Hs.509140
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013448.2 NP_038476.2 19, 20 3573
NM_182648.1 NP_872589.1 18, 19 3477
XM_011536374.1 XP_011534676.1 18, 19 2872
XM_011536376.2 XP_011534678.2 9, 10 1700
XM_017020942.1 XP_016876431.1 8, 9 1603
GenBank mRNA AB032252.1 - 18, 19 3474
AB209255.1 - 10, 11 1824
AF213467.1 - 19, 20 3171
AF221130.1 - 18, 19 3005
AL050089.1 - 2, 3 770

View Full Product Details