Gene Transcripts
- Gene Symbol
- AASDHPPT
- Entrez Gene ID
- 60496
- Gene Name
- aminoadipate-semialdehyde dehydrogenase-phosphopantetheinyl transferase
- Gene Aliases
- AASD-PPT, ACPS, CGI-80, LYS2, LYS5
- Chromosome Location
- Chr. 11: 106077565 - 106098692 on Build GRCh38
- UniGene ID
- Hs.524009
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_015423.2 |
NP_056238.2
|
5, 6 |
916 |
|
GenBank mRNA |
AF136978.1 |
- |
5, 6 |
938 |
|
AF151057.1 |
- |
5, 6 |
936 |
|
AF151838.1 |
- |
5, 6 |
936 |
|
AF201943.1 |
- |
5, 6 |
885 |
|
AF302110.1 |
- |
5, 6 |
936 |
|
AK293362.1 |
- |
4, 5 |
677 |
|
AK312529.1 |
- |
5, 6 |
820 |
|
AL050073.1 |
- |
4, 5 |
477 |
|
BC015470.1 |
- |
5, 6 |
807 |
3914010 |
BC016728.1 |
- |
5, 6 |
836 |
4081713 |
BX537665.1 |
- |
1, 2 |
3194 |
|
View Full Product Details