Gene Transcripts
- Gene Symbol
- NUP54
- Entrez Gene ID
- 53371
- Gene Name
- nucleoporin 54
- Gene Aliases
- Chromosome Location
- Chr. 4: 76114659 - 76148515 on Build GRCh38
- UniGene ID
- Hs.430435
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001278603.1 |
NP_001265532.1
|
3, 4 |
527 |
|
NM_017426.3 |
NP_059122.2
|
4, 5 |
671 |
|
XM_006714237.1 |
XP_006714300.1
|
5, 6 |
648 |
|
XM_011532033.2 |
XP_011530335.1
|
4, 5 |
614 |
|
XM_011532034.2 |
XP_011530336.1
|
3, 4 |
914 |
|
XM_017008294.1 |
XP_016863783.1
|
4, 5 |
1058 |
|
GenBank mRNA |
AF157322.1 |
- |
4, 5 |
552 |
|
AK001517.1 |
- |
4, 5 |
553 |
|
AK222752.1 |
- |
4, 5 |
558 |
|
AK223352.1 |
- |
4, 5 |
554 |
|
AK300036.1 |
- |
3, 4 |
409 |
|
AK315160.1 |
- |
4, 5 |
551 |
|
BC012559.1 |
- |
4, 5 |
568 |
|
View Full Product Details