Gene Transcripts

Gene Symbol
MBTD1
Entrez Gene ID
54799
Gene Name
mbt domain containing 1
Gene Aliases
SA49P01
Chromosome Location
Chr. 17: 51177419 - 51260511 on Build GRCh38
UniGene ID
Hs.656803
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017643.2 NP_060113.2 16 2086
XM_005257467.4 XP_005257524.1 15 2031
XM_005257468.4 XP_005257525.1 15 2024
XM_011524922.2 XP_011523224.1 15 2033
XM_011524923.2 XP_011523225.1 15 2030
XM_011524924.2 XP_011523226.1 15 2026
XM_011524925.1 XP_011523227.1 16 2147
XM_011524926.2 XP_011523228.1 16 2251
XM_011524928.2 XP_011523230.1 15 2188
XM_011524929.2 XP_011523231.1 16 2102
XM_011524930.1 XP_011523232.1 15 1824
XM_011524931.2 XP_011523233.1 14 1658
XM_011524933.2 XP_011523235.1 17 2208
XM_017024782.1 XP_016880271.1 16 2398
GenBank mRNA AK000062.1 - 11 1919
CN310792.1 - 2 118

View Full Product Details