Gene Transcripts

Gene Symbol
SLCO1B3
Entrez Gene ID
28234
Gene Name
solute carrier organic anion transporter family member 1B3
Gene Aliases
HBLRR, LST-2, LST-3TM13, LST3, OATP-8, OATP1B3, OATP8, SLC21A8
Chromosome Location
Chr. 12: 20810704 - 20916911 on Build GRCh38
UniGene ID
Hs.504966
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019844.3 NP_062818.1 16 2649
GenBank mRNA AJ251506.1 - 14 2451
AK055874.1 - 16 2627

View Full Product Details