Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_020750.2 | NP_065801.1 | 13 | 1562 | |
GenBank mRNA | AB033117.2 | - | 13 | 1501 | |
AF271159.1 | - | 13 | 1351 | ||
AF298880.1 | - | 13 | 1414 | ||
AK023431.1 | - | 3 | 154 | ||
AK056619.1 | - | 9 | 887 | ||
AK291452.1 | - | 13 | 1525 | ||
AL834207.1 | - | 13 | 1471 | ||
BC009969.2 | - | 6 | 526 | ||
BC062635.1 | - | 13 | 1528 | 5492046 |