Gene Transcripts

Gene Symbol
HOMEZ
Entrez Gene ID
57594
Gene Name
homeobox and leucine zipper encoding
Gene Aliases
KIAA1443
Chromosome Location
Chr. 14: 23273635 - 23286100 on Build GRCh38
UniGene ID
Hs.632332
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020834.2 NP_065885.2 2 981
GenBank mRNA AB037864.1 - 2 907
AF463523.1 - 2 905
AK296927.1 - 2 941
AK301849.1 - 2 827
AK302788.1 - 2 841
BC130392.1 - 1 806 40146423

View Full Product Details