Gene Transcripts
- Gene Symbol
- SLC13A1
- Entrez Gene ID
- 6561
- Gene Name
- solute carrier family 13 member 1
- Gene Aliases
- NAS1, NaSi-1
- Chromosome Location
- Chr. 7: 123113490 - 123201836 on Build GRCh38
- UniGene ID
- Hs.489849
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001324400.1 |
NP_001311329.1
|
6, 7 |
799 |
|
NM_022444.3 |
NP_071889.2
|
5, 6 |
643 |
|
XM_011516515.2 |
XP_011514817.1
|
5, 6 |
647 |
|
XM_011516516.2 |
XP_011514818.1
|
5, 6 |
647 |
|
XM_011516517.2 |
XP_011514819.1
|
6, 7 |
766 |
|
XM_011516518.2 |
XP_011514820.1
|
5, 6 |
647 |
|
XM_011516519.2 |
XP_011514821.1
|
5, 6 |
647 |
|
XM_017012554.1 |
XP_016868043.1
|
4, 5 |
533 |
|
XM_017012555.1 |
XP_016868044.1
|
5, 6 |
647 |
|
GenBank mRNA |
AF260824.1 |
- |
5, 6 |
643 |
|
AK026413.1 |
- |
5, 6 |
641 |
|
AK314537.1 |
- |
5, 6 |
641 |
|
BC111775.1 |
- |
6, 7 |
799 |
|
View Full Product Details