Gene Transcripts

Gene Symbol
CPNE4
Entrez Gene ID
131034
Gene Name
copine 4
Gene Aliases
COPN4, CPN4
Chromosome Location
Chr. 3: 131533560 - 132285702 on Build GRCh38
UniGene ID
Hs.199877
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001289112.1 NP_001276041.1 15 1739
NM_130808.2 NP_570720.1 15 2367
NM_153429.1 NP_702907.1 16 1727
XM_011512406.2 XP_011510708.1 14 1514
XM_011512407.2 XP_011510709.1 14 1771
XM_011512408.2 XP_011510710.1 14 2899
XM_017005693.1 XP_016861182.1 18 3120
XM_017005695.1 XP_016861184.1 17 1990
XM_017005696.1 XP_016861185.1 9 884
GenBank mRNA AF465770.1 - 18 2067
AF465771.1 - 16 1727
AK056175.1 - 15 1839
AK128117.1 - 19 3617
AK295557.1 - 15 1654
BC014396.1 - 15 1925 3162811
BC028613.1 - 1 492
BC075046.2 - 15 1516 30915576

View Full Product Details