Gene Transcripts

Gene Symbol
PRRC1
Entrez Gene ID
133619
Gene Name
proline rich coiled-coil 1
Gene Aliases
Chromosome Location
Chr. 5: 127517609 - 127555089 on Build GRCh38
UniGene ID
Hs.483259
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001286808.1 NP_001273737.1 3, 4 689
NM_130809.4 NP_570721.1 3, 4 689
XM_006714529.3 XP_006714592.1 3, 4 897
GenBank mRNA AJ427339.1 - 3, 4 642
AJ441110.3 - 3, 4 642
AJ496322.1 - 3, 4 642
AJ515158.1 - 3, 4 642
AJ515428.1 - 3, 4 642
AJ515429.1 - 3, 4 642
AK057437.1 - 3, 4 626
AK090486.1 - 2, 3 514
AL831963.1 - 3, 4 651
AL832056.1 - 3, 4 652
AL832600.2 - 3, 4 648
BC017066.1 - 3, 4 626 3913311

View Full Product Details