Gene Transcripts

Gene Symbol
GALM
Entrez Gene ID
130589
Gene Name
galactose mutarotase
Gene Aliases
BLOCK25, GLAT, HEL-S-63p, IBD1
Chromosome Location
Chr. 2: 38665910 - 38734767 on Build GRCh38
UniGene ID
Hs.435012
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138801.2 NP_620156.1 7 1805
GenBank mRNA BC014916.1 - 7 1587
BC019263.1 - 7 1618 3502667
EU794611.1 - 7 1587

View Full Product Details