Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_145914.2 | NP_666019.1 | 3 | 713 | |
XM_005250568.4 | XP_005250625.1 | 4 | 1855 | ||
XM_006716113.3 | XP_006716176.1 | 3 | 1694 | ||
XM_017012584.1 | XP_016868073.1 | 4 | 1819 | ||
XM_017012585.1 | XP_016868074.1 | 3 | 737 | ||
GenBank mRNA | AF155100.1 | - | 2 | 388 | |
AK292267.1 | - | 3 | 733 | ||
AK301871.1 | - | 5 | 764 | ||
AK313884.1 | - | 3 | 731 | ||
AL136865.1 | - | 2 | 666 | ||
BC047309.1 | - | 3 | 706 | 5111890 | |
BI764964.1 | - | 3 | 712 | ||
CR749301.1 | - | 4 | 597 |