Gene Transcripts

Gene Symbol
LINGO2
Entrez Gene ID
158038
Gene Name
leucine rich repeat and Ig domain containing 2
Gene Aliases
LERN3, LRRN6C
Chromosome Location
Chr. 9: 27937617 - 29213981 on Build GRCh38
UniGene ID
Hs.444665
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001258282.1 NP_001245211.1 7 2082
NM_152570.2 NP_689783.1 7 2162
XM_011517719.1 XP_011516021.1 6 3030
XM_011517724.2 XP_011516026.1 5 6049
XM_011517728.2 XP_011516030.1 2 5828
XM_017014303.1 XP_016869792.1 11 5933
XM_017014304.1 XP_016869793.1 4 8453
XM_017014305.1 XP_016869794.1 5 8036
XM_017014306.1 XP_016869795.1 3 5197
XM_017014307.1 XP_016869796.1 2 2670
GenBank mRNA AK056372.1 - 7 2162
AK172834.1 - 1 1162
AK290972.1 - 2 1693
AY358114.1 - 2 1716
BC137514.1 - 7 2022 9021514
BC144676.1 - 7 2022 9053210

View Full Product Details