Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_002166.4 | NP_002157.2 | 1 | 365 | |
GenBank mRNA | AK222682.1 | - | 1 | 292 | |
AK311988.1 | - | 1 | 292 | ||
AY634687.1 | - | 1 | 301 | ||
BC030639.2 | - | 1 | 292 | 4820416 | |
D13891.1 | - | 1 | 278 | ||
M97796.1 | - | 1 | 290 |