Gene Transcripts

Gene Symbol
KSR2
Entrez Gene ID
283455
Gene Name
kinase suppressor of ras 2
Gene Aliases
Chromosome Location
Chr. 12: 117453012 - 117968558 on Build GRCh38
UniGene ID
Hs.375836
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173598.4 NP_775869.3 8, 9 1366
XM_011538225.2 XP_011536527.1 8, 9 2160
XM_011538226.2 XP_011536528.1 8, 9 1701
XM_011538228.2 XP_011536530.1 6, 7 525
XM_011538229.2 XP_011536531.1 8, 9 1701
XM_017019208.1 XP_016874697.1 8, 9 1701
XM_017019209.1 XP_016874698.1 8, 9 1701
XM_017019210.1 XP_016874699.1 3, 4 151
GenBank mRNA AK098831.1 - 6, 7 544
AY345972.1 - 6, 7 1035
BC107106.1 - 6, 7 1030
BC107107.1 - 6, 7 1030
BC127603.1 - 6, 7 1035 40019272

View Full Product Details