Gene Transcripts

Gene Symbol
MGAT3
Entrez Gene ID
4248
Gene Name
mannosyl (beta-1,4-)-glycoprotein beta-1,4-N-acetylglucosaminyltransferase
Gene Aliases
GNT-III, GNT3
Chromosome Location
Chr. 22: 39447166 - 39492194 on Build GRCh38
UniGene ID
Hs.276808
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001098270.1 NP_001091740.1 1 2183
NM_002409.4 NP_002400.3 2 2298
XM_005261608.4 XP_005261665.1 3 2333
XM_005261609.2 XP_005261666.1 2 2439
XM_006724252.2 XP_006724315.1 2 2143
XM_011530184.2 XP_011528486.1 2 2171
XM_011530185.2 XP_011528487.1 2 3522
XM_011530186.2 XP_011528488.1 2 5168
XM_011530187.2 XP_011528489.1 2 2755
XM_011530188.2 XP_011528490.1 2 2167
XM_011530189.2 XP_011528491.1 2 2088
XM_011530190.2 XP_011528492.1 2 5697
XM_011530191.2 XP_011528493.1 2 2192
XM_011530192.2 XP_011528494.1 2 2084
XM_011530194.2 XP_011528496.1 2 2084
XM_017028802.1 XP_016884291.1 2 4016
GenBank mRNA D13789.1 - 1 2059
DA519457.1 - 1 223

View Full Product Details