Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001323316.1 | NP_001310245.1 | 8 | 1148 | |
NM_001323317.1 | NP_001310246.1 | 6 | 922 | ||
NM_001323318.1 | NP_001310247.1 | 6 | 985 | ||
NM_004613.3 | NP_004604.2 | 7 | 1165 | ||
NM_198951.2 | NP_945189.1 | 7 | 1165 | ||
XM_011529028.1 | XP_011527330.1 | 8 | 1110 | ||
GenBank mRNA | AK058031.1 | - | 7 | 933 | |
AK291714.1 | - | 7 | 1006 | ||
AK295775.1 | - | 6 | 763 | ||
AK300292.1 | - | 6 | 826 | ||
AK314618.1 | - | 7 | 1006 | ||
AY675221.1 | - | 6 | 934 | ||
BC003551.1 | - | 7 | 1018 | 3544757 | |
EU794665.1 | - | 7 | 1034 | ||
M55153.1 | - | 7 | 1069 | ||
M98478.1 | - | 7 | 1066 |