Gene Transcripts

Gene Symbol
ALG6
Entrez Gene ID
29929
Gene Name
ALG6, alpha-1,3-glucosyltransferase
Gene Aliases
CDG1C
Chromosome Location
Chr. 1: 63367590 - 63438562 on Build GRCh38
UniGene ID
Hs.258501
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013339.3 NP_037471.2 4 542
GenBank mRNA AF063604.2 - 4 497
AF102851.1 - 4 475
AK022700.1 - 4 480
AU280221.1 - 4 505
BC001253.1 - 4 474 3452687
BI819904.1 - 4 544

View Full Product Details