Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_002795.3 | NP_002786.2 | 3 | 372 | |
NR_104194.1 | 3 | 372 | |||
NR_104195.1 | 3 | 372 | |||
GenBank mRNA | BC013008.2 | - | 3 | 335 | |
BQ428285.1 | - | 3 | 332 | ||
CN412852.1 | - | 3 | 276 | ||
D26598.1 | - | 3 | 275 |