Gene Transcripts

Gene Symbol
PMPCB
Entrez Gene ID
9512
Gene Name
peptidase, mitochondrial processing beta subunit
Gene Aliases
Beta-MPP, MPP11, MPPB, MPPP52, P-52
Chromosome Location
Chr. 7: 103297412 - 103329902 on Build GRCh38
UniGene ID
Hs.184211
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004279.2 NP_004270.2 4 453
XM_005250717.3 XP_005250774.1 4 467
XM_006716181.3 XP_006716244.1 4 466
GenBank mRNA AF054182.1 - 4 432
AK000994.1 - 4 435
AK098616.1 - 4 423
AK289864.1 - 4 435
AK297354.1 - 4 818
AK309928.1 - 423
BC014079.2 - 4 393
BP311561.1 - 4 451

View Full Product Details